missing translation for 'onlineSavingsMsg'
Learn More

Rauwolscine hydrochloride, MedChemExpress

Product Code. 16465956
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465956 100 mg 100mg
16461507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465956 Supplier Medchem Express Supplier No. HY12710A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rauwolscine hydrochloride is a potent and specific α2 adrenergic receptor antagonist with a Ki of 12 nM.

Rauwolscine hydrochloride is a potent and specific α2 adrenergic receptor antagonist with a Ki of 12 nM.

Specifications

Chemical Name or Material Rauwolscine hydrochloride
CAS 6211-32-1
Color White
Purity Grade Notes Research
Molecular Formula C21H27ClN2O3
Quantity 100 mg
Synonym α-Yohimbine hydrochloride Corynanthidine hydrochloride Isoyohimbine hydrochloride
Solubility Information DMSO : 6 mg/mL (15.35 mM; Need ultrasonic) ∣H2O : 5 mg/mL (12.79 mM; Need ultrasonic)
SMILES [H]Cl.[H][C@]12C(NC3=C4C=CC=C3)=C4CCN1C[C@@]5([H])CC[C@H](O)[C@@H](C(OC)=O)[C@]5(C2)[H]
Molecular Weight (g/mol) 390.9
Formula Weight 390.9
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.