missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rattin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14359
This item is not returnable.
View return policy
Description
PA1-4359 detects Rattin (Humanin-like) from rat samples. PA1-4359 has been successfully used in Western blot procedures. PA1-4359 immunizing peptide corresponds to amino acid residues 20-35 from rat Rattin protein.
A novel rat gene, Rattin, encodes a peptide homologous to Humanin, a secreted peptide that specifically protects against neuronal cell death induced by beta-amyloid peptide (Ab) or by mutations causing early-onset familial Alzheimer's disease. Rattin is the prototype of a novel class of peptides, phylogenetically related to Humanin, endowed with protective activity not only against Ab but also toward excitotoxic neuronal death. The rat gene, Rattin, encodes a peptide of 38 residues (15 residues longer than Humanin) showing 73% identity in the conserved region to Humanin.
Specifications
| Rattin | |
| Polyclonal | |
| Unconjugated | |
| Humanin homolog | |
| Synthetic Peptide: V(20) K R L E S P N K T R R P Y G A(35). | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto