missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rattin Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-246G
This item is not returnable.
View return policy
Description
Rattin Polyclonal antibody specifically detects Rattin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rattin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| Humanin-like in rat, rattin humanin-like | |
| A synthetic peptide mapping to residues 20-35 [VKRLESPNKTRRPYGA] of the Rattin protein. | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction