missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ratchet Nut v3.0 Blue

Product Code. 11421535 Shop All MP Biomedicals Products
Click to view available options
Packaging Quantity:
1 Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11421535

Brand: MP Biomedicals™ 116005510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Ratchet Nut v3.0 Blue is a FastPrep-24™ part/accessory.

  • Securing knob used to maintain the tube holder adapters on the central axis or 3 step assembly of the FastPrep-24™ instruments.
  • Due to its high torque, it is particularly recommended for the heaviest adapters and especially for metal tube holders.
  • Can be used on: FastPrep-24™ Classic or FastPrep-24™ 5G
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ratchet Nut
For Use With (Equipment) FastPrep-24™ Classic, FastPrep-24™ 5G
Packaging Quantity 1 Pk.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.