missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS Ratchet Nut Assembly for FP-96
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-96
Unit Size:
Each
Description
- Ratchet Nut Assembly for FP-96.
- Locking system.
- FastPrep-96 parts and accessories.
Specifications
Specifications
| Product Type | Ratchet Nut |
| For Use With (Equipment) | FastPrep-96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction