missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat UNC5H2/UNC5B Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1006
This item is not returnable.
View return policy
Description
UNC5H2/UNC5B Polyclonal specifically detects UNC5H2/UNC5B in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UNC5H2/UNC5B | |
| Polyclonal | |
| Unconjugated | |
| O08722 | |
| UNC5B | |
| Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722 | |
| 100 μg | |
| Primary | |
| Detects rat UNC5H2 in direct ELISAs and Western blots. | |
| Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| p53RDL1, p53-regulated receptor for death and life protein 1, Protein unc-5 homolog 2, Protein unc-5 homolog B, transmembrane receptor Unc5H2, unc5 (C.elegans homolog) b, unc-5 homolog 2, unc-5 homolog B (C. elegans), UNC5B, UNC5H2netrin receptor UNC5B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 219699 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction