missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNF alpha Rat ELISA Kit
For the quantitative detection of rat TNF-alpha.
£451.00 - £2400.00
Specifications
Accession Number | P16599 |
---|---|
Conjugate | Biotin |
Product Type | ELISA Kit |
Sample Type | Serum, Supernatant |
Target Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15502057
|
Invitrogen™
BMS622TEN |
10 x 96 Tests |
£2400.00
Pack of 10 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
13427093
|
Invitrogen™
BMS622 |
96 Tests |
£451.00
1 set |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
The Rat Tumor Necrosis Factor alpha(Rt TNFα) ELISA quantitates Rt TNFα in rat serum or cell culture medium. The assay will exclusively recognize both natural and recombinant rat TNFα. Principle of the method The Rat TNF&alph
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P16599 | |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
10% | |
HRP | |
Serum, 50 μL; Supernatant, 50 μL | |
Cachectin, Tumor necrosis factor alpha, DIF | |
1 hr. 20 min. |
Biotin | |
Serum, Supernatant | |
ELISA | |
24835 | |
5% | |
RUO | |
2°C to 8°C | |
Rat | |
3 hr. 10 min. |
For Research Use Only