missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat TIMP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £478.00
Specifications
| Antigen | TIMP-1 |
|---|---|
| Clone | 150906 |
| Dilution | Immunoprecipitation 25 ug/mL |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701968
![]() |
R&D Systems
MAB5801-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18151791
![]() |
R&D Systems
MAB5801 |
500 μg |
£478.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-1 Monoclonal specifically detects TIMP-1 in Rat samples. It is validated for Immunoprecipitation.Specifications
| TIMP-1 | |
| Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| TIMP1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 150906 | |
| Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Rat | |
| P30120 | |
| 7076 | |
| Mouse myeloma cell line NS0-derived recombinant rat TIMP-1 Cys24-Ala217 Accession # P30120 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat TIMP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TIMP-1, recombinant mouse TIMP-1, rhTIMP-2, or rhTIMP-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title