missing translation for 'onlineSavingsMsg'
Learn More

Rat TIM-1/KIM-1/HAVCR Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 2 publications

£167.00 - £1535.00

Specifications

Antigen TIM-1/KIM-1/HAVCR
Clone 511227
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18718859
GREENER_CHOICE
View Documents
R&D Systems
MAB3689-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18703349
GREENER_CHOICE
View Documents
R&D Systems
MAB3689-100
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18702017
GREENER_CHOICE
View Documents
R&D Systems
MAB3689-500
500 μg
£1535.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

TIM-1/KIM-1/HAVCR
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Rat
O54947
26762
Mouse Myeloma cell line NS0-derived recombinant rat TIM-1/KIM-1/HAVCR Ser18-Val238 Accession # O54947
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat TIM-1/KIM-1/HAVCR in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) TIM-1, rmTIM-3, rmTIM-4, recombinant human (rh) TIM-1, rhTIM-4, or rhNKp44 is observed.
511227
ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
HAVCR1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.