missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Synaptophysin (aa 253-272) Synthetic Peptide

Product Code. 11564900
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564900 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564900 Supplier Invitrogen™ Supplier No. PEP279

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Rat Synaptophysin (residues 253-272) Synthetic Peptide, PEP-279, from Invitrogen.

Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96. PEP-279 is a 20 amino acid synthetic peptide whose sequence corresponds to residues 253-272 of rat Synaptophysin. The sequence of this peptide is (amino to carboxy terminus): G (253)-P-G-G-Y-G-P-Q-D-S-Y-G-P-Q-G-G-Y-Q-P-D (272).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, Synaptophysin, catalog &Num; PA1-1043. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 24804
Species Rat
Purification Method purified
Accession Number P07825
Common Name Rat Synaptophysin (aa 253-272)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Syp
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence G(253) - P - G - G - Y - G - P - Q - D - S - Y - G - P - Q - G - G - Y - Q - P - D(272)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.