missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rat Skin Whole Tissue Lysate (Adult Whole Normal)
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
The Rat Skin Whole Tissue Lysate (Adult Whole Normal) is derived from rat. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Skin |
| Quantity | 0.1 mg |
| Storage Buffer | Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. |
| Test Specificity | Rat skin tissue from outbred Sprague Dawley Albino rat |
| Host Species | Rat |
| Concentration | 2 mg/mL |
| Content And Storage | Store at −80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction