missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat ROBO1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB17491
Additional Details : Weight : 0.00970kg
Description
ROBO1 Monoclonal specifically detects ROBO1 in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
ROBO1 | |
Monoclonal | |
LYOPH | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
O55005 | |
ROBO1 | |
Mouse myeloma cell line NS0-derived recombinant rat ROBO1 Lys19-Ile560 Accession # O55005 | |
500 μg | |
Primary | |
Detects rat ROBO1 in direct ELISAs and Western blots. | |
Rat | |
Purified |
ELISA | |
243706 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6091 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only