missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Reg3B Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1996-SP
This item is not returnable.
View return policy
Description
Reg3B Polyclonal specifically detects Reg3B in Rat samples. It is validated for Western Blot.
Specifications
| Reg3B | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PAP1, Reg3B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 18489 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P25031 | |
| Reg3b | |
| E. coli-derived recombinant rat Reg3B Glu27-Gly175 Accession # P25031 | |
| 25 μg | |
| Primary | |
| Detects rat Reg3B in direct ELISAs and Western blots. In direct ELISAs, approxmately 75% cross-reactivity with recombinant mouse (rm) Reg3B, and approximately 20% cross-reactivity with rmReg3G is observed, and less than 2% cross-reactivity with recombinant human (rh) Reg3A, rmReg1, rmReg2, rhReg4, rhReg1A, rmReg3A, and rmReg3D is observed. In Western blots, approximately 40% cross-reactivity with rmReg3B is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction