missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Rat Recombinant M-CSF

Product Code. p-7217359
Encompass_Preferred
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 17189601

Brand: STEMCELL Technologies 78117

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Macrophage colony-stimulating factor

  • Macrophage colony-stimulating factor (M-CSF) is a homodimeric cytokine that belongs to receptor tyrosine kinase subclass III of the receptor tyrosine kinase (RTK) family
  • M-CSF acts on a CSF-1 receptor tyrosine kinase, which initiates signaling cascades to support cell proliferation and differentiation (Hamilton)
  • M-CSF is produced by endothelial cells, osteoblasts, and during pregnancy by the uterine epithelial cells (Ryan)
  • M-CSF synergizes with other factors to support proliferation and differentiation of multipotent hematopoietic progenitor cells, and regulates proliferation and differentiation of the mononuclear phagocyte progenitor cells to monocytes and macrophages
  • It also supports survival, proliferation, and function of the differentiated macrophages and regulates differentiation of mononuclear phagocytes to osteoclasts (Pixley & Stanley)
  • M-CSF plays an important role in the implantation of the embryo and early development (Makrigiannakis et al.)

Specifications

Name CSF-1, Macrophage colony-stimulating factor
Quantity 5 μg
Cell Type Hematopoietic Stem and Progenitor Cells; Monocytes
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.