missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Rat Recombinant M-CSF
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
Description
- Macrophage colony-stimulating factor (M-CSF) is a homodimeric cytokine that belongs to receptor tyrosine kinase subclass III of the receptor tyrosine kinase (RTK) family
- M-CSF acts on a CSF-1 receptor tyrosine kinase, which initiates signaling cascades to support cell proliferation and differentiation (Hamilton)
- M-CSF is produced by endothelial cells, osteoblasts, and during pregnancy by the uterine epithelial cells (Ryan)
- M-CSF synergizes with other factors to support proliferation and differentiation of multipotent hematopoietic progenitor cells, and regulates proliferation and differentiation of the mononuclear phagocyte progenitor cells to monocytes and macrophages
- It also supports survival, proliferation, and function of the differentiated macrophages and regulates differentiation of mononuclear phagocytes to osteoclasts (Pixley & Stanley)
- M-CSF plays an important role in the implantation of the embryo and early development (Makrigiannakis et al.)
Specifications
Specifications
| Name | CSF-1, Macrophage colony-stimulating factor |
| Quantity | 25 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Monocytes |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction