missing translation for 'onlineSavingsMsg'
Learn More

Rat Pure-Blot anti-Mouse IgG (H+L) Secondary Antibody (eB144), Biotin, Novus Biologicals™

Product Code. 18348801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348801 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348801 Supplier Novus Biologicals Supplier No. NBP311660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone eB144
Conjugate Biotin
Dilution Western Blot 1:000, Immunoprecipitation
Formulation 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 (Proprietary Stabilizer)
Gene Alias IP Detection Reagent
Host Species Rat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.