missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat Prolactin Recombinant Protein, PeproTech®

Product Code. 17854063
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17854063

Brand: Gibco™ 4002750UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat Prolactin is a 22.6 kDa globular protein containing 198 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01237
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 24683
Molecular Weight (g/mol) 22.6 kDa
Name Rat Prolactin
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL
Common Name Prolactin
Gene Symbol PRL
Biological Activity Determined by its ability to induce the proliferation of rat Nb2-11 cells in the concentration range of 0.1-1.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLPVCSGGDC QTPLPELFDR VVMLSHYIHT LYTDMFIEFD KQYVQDREFI AKAINDCPTS SLATPEDKEQ AQKVPPEVLL NLILSLVHSW NDPLFQLITG LGGIHEAPDA IISRAKEIEE QNKRLLEGIE KIISQAYPEA KGNEIYLVWS QLPSLQGVDE ESKDLAFYNN IRCLRRDSHK VDNYLKFLRC QIVHKNNC
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.