missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Pentraxin 2/SAP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1895-SP
This item is not returnable.
View return policy
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Western Blot.
Specifications
| Pentraxin 2/SAP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P23680 | |
| APCS | |
| Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 (predicted) Accession # P23680 | |
| 25 μg | |
| Primary | |
| Detects rat Pentraxin 2/SAP in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2 and 25% cross-reactivity with recombinant mouse (rm) Pentraxin 2 is observed. In direct ELISAs, no cross-reactivity with rhPentraxin 2 or rmPentraxin 2 is observed. | |
| Rat | |
| Purified |
| Western Blot | |
| 273912 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 325 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction