missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1055
This item is not returnable.
View return policy
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Rat samples. It is validated for Western Blot.
Specifications
| PDGF-AA | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAB59693 | |
| Pdgfa | |
| E. coli-derived recombinant rat PDGF-AA Ser87-Arg196 Accession # AAB59693 | |
| 500 μg | |
| Primary | |
| Detects rat PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, 5-35% cross-reactivity with recombinant rat (rr) PDGF-AB, recombinant human (rh) PDGF-AA, and rhPDGF-AB is observed and no cross-reactivity with rrPDGF-BB, rhPDGF-BB, or rhPDGF-CC is observed. | |
| Rat | |
| Purified |
| Western Blot | |
| 184805 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5154 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction