missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat OX40 Ligand/TNFSF4 Protein

Product Code. 18707536
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18707536

Brand: R&D Systems™ 8640OX025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.05-0.25 μg/mL in the presence of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z2P3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 7292
Molecular Weight (g/mol) M.W. (Observed): 17-31 kDa, reducing conditions; M.W. (theoretical): 18 kDa
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived rat OX40 Ligand/TNFSF4 protein Gln49-Leu199, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Common Name OX40 Ligand/TNFSF4
Gene Symbol Tnfsf4
Biological Activity Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is typically 0.05-0.25μg/mL in the presence of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody.
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS.
Expression System Mouse myeloma cell line, NS0-derived Gln49-Leu199, with an N-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein OX40 Ligand/TNFSF4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.