missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB86291
This item is not returnable.
View return policy
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.
Specifications
| mu Opioid R/OPRM1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4988 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 1126D | |
| Western Blot 0.5 ug/mL | |
| P33535 | |
| OPRM1 | |
| Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535 | |
| 100 μg | |
| Primary | |
| Detects rat μ Opioid R/OPRM1 in direct ELISAs. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction