Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6866-SP
Product Code. 18781099
  • £171.00 / 25µg
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mu Opioid R/OPRM1
Monoclonal
LYOPH
Western Blot 1 ug/mL
P33535
OPRM1
Synthetic peptide corresponding to His385-Pro398 of rat μ Opioid R/OPRM1 Accession # P33535
25 μg
Primary
Detects rat μ Opioid R/OPRM1 in direct ELISAs.
Rat
Purified
Western Blot
677014
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4988
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a