Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB8629
Product Code. 18717749
  • £362.00 / 100µL
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mu Opioid R/OPRM1
Monoclonal
Unconjugated
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1
Rabbit
Protein A or G purified from cell culture supernatant
RUO
4988
Rat
Supernatant
Immunohistochemistry
1126B
Immunohistochemistry 1:125 dilution
P33535
OPRM1
Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535
100 μg
Primary
Detects rat μ Opioid R/OPRM1 in direct ELISAs.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.