missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Nogo-A aa 2-172 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF3098-SP
This item is not returnable.
View return policy
Description
Nogo-A Polyclonal specifically detects Nogo-A in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nogo-A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Rtn4 | |
| E. coli-derived recombinant rat Nogo-A Glu2-Val172 Accession # Q9JK11 | |
| 25 μg | |
| Primary | |
| Detects rat Nogo-A in direct ELISAs and Western blots. | |
| Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9JK11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57142 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction