missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Nogo-A aa 2-172 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF3098
This item is not returnable.
View return policy
Description
Nogo-A Polyclonal specifically detects Nogo-A in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nogo-A | |
| Polyclonal | |
| Unconjugated | |
| Q9JK11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57142 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Rtn4 | |
| E. coli-derived recombinant rat Nogo-A Glu2-Val172 Accession # Q9JK11 | |
| 100 μg | |
| Primary | |
| Detects rat Nogo-A in direct ELISAs and Western blots. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction