missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Neuropilin-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5661V-100UG
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Rat samples. It is validated for Flow Cytometry.
Specifications
| Neuropilin-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9QWJ9 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1 Met1-Asp854 (Lys811Arg, Pro812-Gly828 del), predicted Accession # Q9QWJ9 | |
| RUO | |
| 8829 | |
| Rat | |
| IgG1 |
| Flow Cytometry | |
| 130604 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects rat Neuropilin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat Neuropilin-2 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction