missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Neurexophilin-1/NXPH-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4208
This item is not returnable.
View return policy
Description
Neurexophilin-1 Polyclonal specifically detects Neurexophilin-1 in Rat samples. It is validated for Western Blot.
Specifications
| Neurexophilin-1 | |
| Polyclonal | |
| Unconjugated | |
| Q63366 | |
| NXPH1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-1/NXPH-1 Ala22-Gly271 Accession # Q63366 | |
| 100 μg | |
| Primary | |
| Detects rat Neurexophilin-1/NXPH-1 in direct ELISAs and Western blots. | |
| Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| neurexophilin 1, neurexophilin-1, Neurexophilin1, NPH1, NXPH1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 30010 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction