missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Multiple Tissue MicroArray (Normal)

Product Code. 18700254 Shop All Bio Techne Products
Change view
Click to view available options
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18700254 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700254 Supplier Novus Biologicals™ Supplier No. NBP252397
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Sprague Dawley or Wistar rat tissues.

TRUSTED_SUSTAINABILITY

Specifications

Description Paraffin Tissue Section Panel - Rat Normal Tissue, Multitissue Set I mounted on positively charged glass slides
Quantity 5 Slides
Species Rat
Tissue Condition Normal
Type Multiple
For Use With (Application) Paraffin Tissue Section Panel - Rat Normal Tissue, Multi-tissue I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.