missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat MMP-8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3245
This item is not returnable.
View return policy
Description
MMP-8 Monoclonal specifically detects MMP-8 in Rat samples. It is validated for Western Blot.
Specifications
| MMP-8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O88766 | |
| MMP8 | |
| Mouse myeloma cell line NS0-derived recombinant rat MMP-8 Leu21-Pro466 Accession # O88766 | |
| 100 μg | |
| Primary | |
| Detects rat MMP-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse MMP-8 and 50% cross-reactivity with recombinant human MMP-8 is observed. | |
| Rat | |
| Purified |
| Western Blot | |
| 400102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4317 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido