missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat MBP ELISA Kit (Colorimetric)

Product Code. 18339410 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339410 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339410 Supplier Novus Biologicals™ Supplier No. NBP281167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This kit recognizes Rat MBP in samples.

No significant cross-reactivity or interference between Rat MBP and analogues was observed.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 4155
Gene Symbol MBP
Host Species Rat
Quantity 1 Kit
Research Discipline Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target MBP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.