missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat MBL-2 ELISA Kit

Product Code. 17115879
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17115879

Brand: Invitrogen™ ERA38RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Rat MBL-2 quantitates rat MBL-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat MBL-2.

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Ab2-001; Ab2-011; COLEC1; Collectin1; Collectin-1; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (C); Mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding protein C; mannose-binding protein C (liver); MBL; Mbl2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A
Assay Range 26 to 6,000 pg/mL
Assay Sensitivity 26 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 64668
Gene Symbol MBL2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MBL2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.