missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Rat Luminex™ Discovery Assay
Click to view available options
Quantity:
10 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
3 Plex
5 Plex
6 Plex
7 Plex
9 Plex
Unit Size:
1 set
Each
Specifications
Specifications
| Quantity | 16 Plex |
| Species | Rat |
| Binding Target | Multiplex Cytokine Assay Kits |
| Content And Storage | Store the unopened kit at 2-8 °C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction