missing translation for 'onlineSavingsMsg'
Learn More

Rat MAG/Siglec-4a Antibody, R&D Systems™

Product Code. 18022342
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022342 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18022342 Supplier R&D Systems Supplier No. AF538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

MAG/Siglec-4a Polyclonal specifically detects MAG/Siglec-4a in Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAG/Siglec-4a
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P07722
Gene Alias GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG
Gene Symbols MAG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat MAG/Siglec-4a Gly20-Pro516 Accession # P07722
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4099
Test Specificity Detects rat MAG/Siglec-4a in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.