missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat liver tissue lysate - mitochondrial extract, 2 mg

Product Code. 32153161 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
2000µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32153161 2000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32153161 Supplier Abcam Supplier No. ab1103462mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat liver cell/tissue lysate.

  • Species: Rat
  • Tissue: liver
  • Form: Liquid

Specifications

For Use With (Application) Western Blot, SDS-PAGE, Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.