missing translation for 'onlineSavingsMsg'
Learn More

Rat liver tissue lysate, Denatured; Abnova

Product Code. 16069595
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16069595 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16069595 Supplier Abnova Supplier No. L038W1.200ug
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non denatured tissue lysate of Rat liver

  • Tissue: Liver
  • Host: Rat
  • Lysis buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
  • Storage buffer: In ready-to-use 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)

Western Blotting

Specifications

For Use With (Application) Western Blot
Tissue Liver
Description Whole tissue lysate (denatured)
Lysis Buffer Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
Preparation Method Tissue lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was adjusted to 3.75 mg/ml, and then mixed with 5X Sample Buffer to become final 3 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly.
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 200 μg
Storage Buffer In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue).
Host Species Rat
Concentration 3 mg/mL
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.