Learn More
Invitrogen™ Rat Leptin Receptor (aa 577-594) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP014
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Leptin Receptor (OB-R) (residues 577-594) Synthetic Peptide, PEP-014, from Invitrogen.
PEP-014 is an 18 amino acid (amino acids 577-594) synthetic peptide whose sequence is from the extracellular domain of the leptin receptor (OB-R). The sequence of this peptide is (amino to carboxy terminus): K-E-I-Q-W-K-T-H-E-V-F-D-A-K-S-K-S-AThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and leptin receptor (OB-R), catalog # PA1-053. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
24536 | |
purified | |
Rat Leptin Receptor (aa 577-594) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
Q62959 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Lepr | |
No preservative | |
50 μg | |
K - E - I - Q - W - K - T - H - E - V - F - D - A - K - S - K - S - A |