missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-9 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4134-SP
This item is not returnable.
View return policy
Description
IL-9 Polyclonal specifically detects IL-9 in Rat samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-9 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3578 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 1.5-7.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_001099217 | |
| IL9 | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-9 Gln19-Ala144 Accession # NP_001099217 | |
| 25 μg | |
| Primary | |
| Detects rat IL-9 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse IL-9 is observed, and approximately 10% cross-reactivity with recombinant human IL-9 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction