missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-7R alpha/CD127 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£248.00 - £457.00
Specifications
| Antigen | IL-7R alpha/CD127 |
|---|---|
| Clone | 717519 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18115011
|
R&D Systems
FAB8484P-100 |
100 Tests |
£457.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713708
![]() |
R&D Systems
FAB8484P-025 |
25 Tests |
£248.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry.Specifications
| IL-7R alpha/CD127 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| IL7R | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 717519 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Rat | |
| NP_001099888 | |
| 3575 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects rat IL-7 R alpha/CD127 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title