missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-7R alpha/CD127 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5607-SP
This item is not returnable.
View return policy
Description
IL-7R alpha/CD127 Polyclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| IL-7R alpha/CD127 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3575 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_001099888 | |
| IL7R | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession # NP_001099888 | |
| 25 μg | |
| Primary | |
| Detects rat IL-7 R alpha/CD127 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse IL-7 R alpha and less than 10% cross-reactivity with recombinant human IL-7 R alpha is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction