missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-18 BPa (NP_445826) VersaClone cDNA

Product Code. 18739908
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18739908 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18739908 Supplier R&D Systems™ Supplier No. RDC2553
  • ONLINE EXCLUSIVE PRICE
    £304.00
    £286.65 / 10µg
    Save £17.35
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Il18bp interleukin 18 binding protein [ Rattus norvegicus (Norway rat) ]
  • The IL-18 BPa/IL18BP insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • Interleukin 18 binding proteins are secreted glycoproteins which antagonize IL-18 activity by binding IL-18 and blocking its biological activity.
  • Alternative splicing of the IL-18 BP gene produces at least four isoforms that differ in affinity for IL-18.
  • The IL-18 BP isoforms a and c each contain one immunoglobulin (Ig)-like C2-type domain while isoforms b and d lack a complete Ig-like domain.
  • IL-18 BPa functions as an inhibitor of the proinflammatory cytokine, IL18.
  • It binds IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production, resulting in reduced Thelper type 1 immune responses.
  • It is constitutively expressed and secreted in mononuclear cells.
  • Elevated level of IL-18 BPa is detected in the intestinal tissues of patients with Crohn's disease.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene rIl18bp
Accession Number NP_445826
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 601 bp
Gene ID (Entrez) 10068
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Rat
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.