Learn More
Invitrogen™ Rat IL-1 beta ELISA Kits
Description
Assay IL-1β (rat interleukin-1 beta) levels in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Rat IL-1 beta ELISA Kits.
Target: Natural and recombinant rat interleukin-1 beta (IL-1β)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 25.6 to 2500pg/mL
Sensitivity: < 12pg/mL
Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3.5 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-2, IL-4, IL-6, IL-10, MCP-1, MIP-1α, or RANTES, or human, mouse or pig IL-1β
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.
Specifications
Specifications
| Assay Range | 25.6 to 2500pg/mL |
| Assay Sensitivity | <12pg/mL |
| Description | Rat IL-1 beta ELISA Kit |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | No significant cross-reaction with rat IFNg, IL-1a, IL-2, IL-4, IL-6, IL-10, MCP-1, MIP-1a, or RANTES, or human, mouse or pig IL-1ß |
| Sufficient For | 96 microplate wells |
| Kit Contents | Lyophilized Recombinant Rat IL-1 beta Standard, 2 vials, TMB Substrate Solution, 12mL, Streptavidin-HRP Concentrate, 75 μL, Stop Solution, 13mL, Sample Diluent, 25mL, HRP Dilution Buffer, 13mL, Biotinylated Antibody Reagent, 13mL, Anti-Rat IL-1 beta Precoated 96-well Strip Plate, 1 ea, Adhesive Plate Covers, 6 ea, 30X Wash Buffer, 50mL |
| Measurement | Absorbance at 450nm minus absorbance at 550nm |
| Precision | Intra-assay CV ≤ 10%; inter-assay CV ≤ 10% |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.