missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Rat IL-1 beta ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Rat
Unit Size:
96 tests
Beschreibung
- The assay recognizes rat IL1-beta. An antibody specific for IL1-beta has been pre-coated onto the microwells.
- The IL-1 beta protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for IL-1 beta is added to detect the captured IL-1 beta protein.
- For signal development, Sterptravidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
- Interleukin-1 is a pro-inflammatory cytokine with multiple biological effects. The IL1 gene family encodes three proteins: IL1 -alpha IL1- beta and their naturally occurring inhibitor IL1-RN. Interleukin 1 beta(IL1- beta), mainly produced by blood monocytes and tissue macrophages, has been implicated in mediating both acute and chronic inflammation.
- IL1-beta is known to be involved in a variety of cellular activities, including cell proliferation, differentiation and apoptosis. IL1-beta is emerging as a key mediator of carcinogenesis that characterizes host-environment interactions.
Spezifikation
Spezifikation
| Accession Number | NP_113700 |
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 12.5 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Rat |
| Gene ID (Entrez) | 24494 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 96TEST Rat IL-1 beta ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur