missing translation for 'onlineSavingsMsg'
Learn More

Rat IgM Isotype Control (eBRM), PE, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 12-4341-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278769

  • £79.75 / 50µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Description: The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control PE has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.
Specifications

Specifications

Rat IgM
eBRM
PE
Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
50 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgM κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.