missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2B PerCP-conjugated Isotype Control, R&D Systems™

Product Code. 18154991
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18154991 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18154991 Supplier R&D Systems Supplier No. IC013C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for Control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2b
Classification Monoclonal
Clone 141945
Conjugate PerCP
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Rat
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as a rat IgG 2B isotype control for flow cytometry applications.
Target Species Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.