missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2b kappa Isotype Control (eB149/10H5), FITC, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 11-4031-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228209

  • £180.00 / 500µg
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2b, kappa, isotype control has been reported for use as a control for immunofluorescent staining, flow cytometric analysis, immunohistochemistry and immunocytochemistry. Applications Tested: This rat IgG2b, kappa, isotype control has been tested by immunofluorescent staining and flow cytometric analysis of mouse splenocyte suspensions and can be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.
Specifications

Specifications

Rat IgG2b kappa
eB149/10H5
FITC
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
500 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.