missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2b kappa Isotype Control (eB149/10H5), Biotin, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 13-4031-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299789

  • £47.64 / 50µg
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: Biotin Rat IgG2b Isotype Control has been reported for use in immunohistochemistry, immunocytochemistry and flow cytometric analysis. Applications Tested: Biotin Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. Use at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: Biotin Rat IgG2b Isotype Control has been reported for use in immunohistochemistry, immunocytochemistry and flow cytometric analysis.Applications Tested: Biotin Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. Use at the same concentration as the experimental antibody.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Rat IgG2b kappa
eB149/10H5
Biotin
IgG2, IgG2b, IgG2bk
Affinity chromatography
RUO
None
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Rat
50 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.