Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), Biotin, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 13-4031-85
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: Biotin Rat IgG2b Isotype Control has been reported for use in immunohistochemistry, immunocytochemistry and flow cytometric analysis. Applications Tested: Biotin Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. Use at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.Specifications
Rat IgG2b kappa | |
eB149/10H5 | |
Biotin | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
500 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.