Learn More
Rat IgG2B Fluorescein-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems IC013F
Product Code. 18164931
  • £280.00 / 200 tests
Description
IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG2b
141945
Control 10 uL/10^6 cells
Rat
Protein A or G purified from hybridoma culture supernatant
Selected for use as a rat IgG 2B isotype control for flow cytometry applications.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
KLH
200 Tests
Rat
IgG2b