Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 570, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 41-4321-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin. Applications Reported: This eBR2a antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This Rat IgG2a Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Rat IgG2a kappa | |
eBR2a | |
eFluor 570 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.