missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 450, eBioscience™, Invitrogen™
Rat Isotype Control
£66.50 - £167.00
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | eFluor 450 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15289175
|
Affymetrix eBioscience
48-4321-80 |
25 μg |
£66.50
25µg |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
13579200
|
Affymetrix eBioscience
48-4321-82 |
100 μg |
£167.00
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG2a Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. . Applications Tested: This Rat IgG2a Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2a kappa | |
0.2 mg/mL | |
eFluor 450 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.